Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
Chromosome Location
Chr.10: 126424997 - 126671038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004298.2 NP_001004298.2 3 - 4 1368 67
XM_005269491.1 XP_005269548.1 3 - 4 1438 67
XM_011539216.1 XP_011537518.1 4 - 5 2096 67
XM_011539217.2 XP_011537519.1 4 - 5 1791 67
XM_011539220.2 XP_011537522.1 4 - 5 1336 67
XM_011539222.2 XP_011537524.1 4 - 5 1458 67
XM_011539223.2 XP_011537525.1 3 - 4 2146 67
XM_011539224.2 XP_011537526.1 3 - 4 1292 67
XM_011539226.1 XP_011537528.1 3 - 4 1291 67
XM_011539227.2 XP_011537529.1 3 - 4 1624 67
GenBank mRNA AK057500.1 - 3 - 4 1368 67
AK297577.1 - 4 - 5 1568 67
BC140898.1 - 3 - 4 1355 67
BC146869.1 - 3 - 4 1355 67

View Full Product Details