The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004298.2 | NP_001004298.2 | 4 - 5 | 1655 | 169 |
XM_005269491.1 | XP_005269548.1 | 4 - 5 | 1725 | 169 | |
XM_005269492.2 | XP_005269549.1 | 2 - 3 | 1018 | 169 | |
XM_011539216.1 | XP_011537518.1 | 5 - 6 | 2383 | 169 | |
XM_011539217.2 | XP_011537519.1 | 5 - 6 | 2078 | 169 | |
XM_011539220.2 | XP_011537522.1 | 5 - 6 | 1623 | 169 | |
XM_011539222.2 | XP_011537524.1 | 5 - 6 | 1745 | 169 | |
XM_011539223.2 | XP_011537525.1 | 4 - 5 | 2433 | 169 | |
XM_011539224.2 | XP_011537526.1 | 4 - 5 | 1579 | 169 | |
XM_011539226.1 | XP_011537528.1 | 4 - 5 | 1578 | 169 | |
XM_011539227.2 | XP_011537529.1 | 4 - 5 | 1911 | 169 | |
XM_011539230.2 | XP_011537532.1 | 3 - 4 | 1700 | 169 | |
XM_011539231.1 | XP_011537533.1 | 4 - 5 | 3710 | 169 | |
XM_011539232.1 | XP_011537534.1 | 4 - 5 | 1377 | 169 | |
XM_011539233.1 | XP_011537535.1 | 3 - 4 | 1140 | 169 | |
GenBank mRNA | AK057500.1 | - | 4 - 5 | 1655 | 169 |
AK297577.1 | - | 5 - 6 | 1855 | 169 | |
BC021140.1 | - | 1 - 2 | 169 | 169 | |
BC140898.1 | - | 4 - 5 | 1642 | 169 | |
BC146869.1 | - | 4 - 5 | 1642 | 169 |