The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003601.3 | NP_003592.3 | 1 - 2 | 638 | 121 |
GenBank mRNA | AB010882.1 | - | 1 - 2 | 378 | 121 |
AK302841.1 | - | 2 - 3 | 273 | 121 | |
AK308424.1 | - | 1 - 2 | 368 | 121 | |
BC023144.1 | - | 1 - 2 | 389 | 121 |