The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257386.1 | NP_001244315.1 | 12 - 13 | 1844 | 124 |
NM_004827.2 | NP_004818.2 | 12 - 13 | 1989 | 124 | |
XM_005263354.3 | XP_005263411.1 | 12 - 13 | 1832 | 124 | |
XM_005263355.3 | XP_005263412.1 | 12 - 13 | 1939 | 124 | |
XM_005263356.3 | XP_005263413.1 | 12 - 13 | 1709 | 124 | |
XM_011532420.2 | XP_011530722.1 | 14 - 15 | 2103 | 124 | |
XM_017008852.1 | XP_016864341.1 | 12 - 13 | 1826 | 124 | |
XM_017008853.1 | XP_016864342.1 | 12 - 13 | 1629 | 124 | |
GenBank mRNA | AB051855.1 | - | 12 - 13 | 1540 | 124 |
AB056867.1 | - | 11 - 12 | 1496 | 124 | |
AF103796.1 | - | 12 - 13 | 1700 | 124 | |
AF463519.1 | - | 11 - 12 | 1504 | 124 | |
AK002040.1 | - | 9 - 10 | 1178 | 124 | |
AK290000.1 | - | 12 - 13 | 1560 | 124 | |
AY288307.1 | - | 9 - 10 | 1154 | 124 | |
AY289766.1 | - | 12 - 13 | 1696 | 124 | |
AY333755.1 | - | 11 - 12 | 1505 | 124 | |
AY333756.1 | - | 11 - 12 | 1505 | 124 | |
BC021281.2 | - | 12 - 13 | 1989 | 124 | |
BC092408.1 | - | 12 - 13 | 1844 | 124 |