The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289002.1 | NP_001275931.1 | 10 - 11 | 1987 | 140 |
NM_001289004.1 | NP_001275933.1 | 9 - 10 | 1801 | 140 | |
NM_001308014.1 | NP_001294943.1 | 5 - 6 | 1228 | 140 | |
NM_173488.4 | NP_775759.3 | 10 - 11 | 1987 | 140 | |
XM_005271874.3 | XP_005271931.1 | 10 - 11 | 5184 | 140 | |
XM_011543147.2 | XP_011541449.1 | 10 - 11 | 5079 | 140 | |
XM_011543148.2 | XP_011541450.1 | 10 - 11 | 4947 | 140 | |
XM_011543150.1 | XP_011541452.1 | 9 - 10 | 1197 | 140 | |
XM_011543153.2 | XP_011541455.1 | 8 - 9 | 1077 | 140 | |
GenBank mRNA | AF505657.1 | - | 10 - 11 | 1958 | 140 |
AK131445.1 | - | 5 - 6 | 1125 | 140 | |
AY273897.1 | - | 10 - 11 | 1950 | 140 | |
BC034976.2 | - | 9 - 10 | 1803 | 140 |