The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289002.1 | NP_001275931.1 | 2 - 3 | 785 | 101 |
NM_173488.4 | NP_775759.3 | 2 - 3 | 785 | 101 | |
XM_005271874.3 | XP_005271931.1 | 2 - 3 | 3982 | 101 | |
XM_011543154.2 | XP_011541456.1 | 2 - 3 | 3982 | 101 | |
GenBank mRNA | AF505657.1 | - | 2 - 3 | 756 | 101 |
AY273897.1 | - | 2 - 3 | 748 | 101 | |
BM554439.1 | - | 2 - 3 | 772 | 101 |