The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005907.3 | NP_005898.2 | 9 - 10 | 1771 | 83 |
XM_005266986.4 | XP_005267043.1 | 9 - 10 | 1627 | 83 | |
XM_011535833.2 | XP_011534135.1 | 8 - 9 | 823 | 83 | |
GenBank mRNA | AK025599.1 | - | 9 - 10 | 1766 | 83 |
AK307383.1 | - | 9 - 10 | 1766 | 83 |