The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005270591.2 | XP_005270648.1 | 13 - 14 | 1697 | 111 |
XM_017000559.1 | XP_016856048.1 | 13 - 14 | 1627 | 111 | |
GenBank mRNA | BC007912.2 | - | 1 - 2 | 77 | 111 |
BC014080.2 | - | 9 - 10 | 1063 | 111 | |
BC016880.1 | - | 7 - 8 | 820 | 111 |