The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032449.2 | NP_115825.1 | 16 - 17 | 2050 | 77 |
XM_005270590.2 | XP_005270647.1 | 16 - 17 | 2136 | 77 | |
XM_005270591.2 | XP_005270648.1 | 16 - 17 | 2118 | 77 | |
XM_005270593.2 | XP_005270650.1 | 12 - 13 | 1872 | 77 | |
XM_017000559.1 | XP_016856048.1 | 16 - 17 | 2048 | 77 | |
GenBank mRNA | AB058739.1 | - | 12 - 13 | 1481 | 77 |
BC007912.2 | - | 4 - 5 | 498 | 77 | |
BC014080.2 | - | 12 - 13 | 1532 | 77 | |
BC016880.1 | - | 10 - 11 | 1241 | 77 | |
BC039308.1 | - | 14 - 15 | 2874 | 77 |