The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178844.3 | NP_849172.2 | 14 - 15 | 3104 | 69 |
NR_075083.2 | 15 - 16 | 3301 | 69 | ||
XM_017023027.1 | XP_016878516.1 | 14 - 15 | 3134 | 69 | |
XM_017023028.1 | XP_016878517.1 | 15 - 16 | 3308 | 69 | |
XM_017023029.1 | XP_016878518.1 | 16 - 17 | 3389 | 69 | |
XM_017023030.1 | XP_016878519.1 | 16 - 17 | 3450 | 69 | |
XM_017023031.1 | XP_016878520.1 | 14 - 15 | 3195 | 69 | |
XM_017023032.1 | XP_016878521.1 | 16 - 17 | 4151 | 69 | |
XM_017023033.1 | XP_016878522.1 | 16 - 17 | 3328 | 69 | |
XM_017023034.1 | XP_016878523.1 | 15 - 16 | 3247 | 69 | |
XM_017023035.1 | XP_016878524.1 | 15 - 16 | 3276 | 69 | |
XM_017023036.1 | XP_016878525.1 | 14 - 15 | 3835 | 69 | |
XM_017023037.1 | XP_016878526.1 | 15 - 16 | 3029 | 69 | |
XM_017023038.1 | XP_016878527.1 | 15 - 16 | 3002 | 69 | |
XM_017023039.1 | XP_016878528.1 | 14 - 15 | 3134 | 69 | |
GenBank mRNA | AF501291.1 | - | 7 - 8 | 581 | 69 |
AK074109.1 | - | 10 - 11 | 982 | 69 | |
AK090431.1 | - | 15 - 16 | 3297 | 69 | |
AY601811.1 | - | 14 - 15 | 3091 | 69 | |
FJ889357.1 | - | 11 - 12 | 2680 | 69 |