The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178844.3 | NP_849172.2 | 2 - 3 | 335 | 85 |
NR_075083.2 | 2 - 3 | 396 | 85 | ||
XM_017023027.1 | XP_016878516.1 | 2 - 3 | 338 | 85 | |
XM_017023028.1 | XP_016878517.1 | 3 - 4 | 512 | 85 | |
XM_017023029.1 | XP_016878518.1 | 4 - 5 | 593 | 85 | |
XM_017023030.1 | XP_016878519.1 | 4 - 5 | 654 | 85 | |
XM_017023031.1 | XP_016878520.1 | 2 - 3 | 399 | 85 | |
XM_017023032.1 | XP_016878521.1 | 4 - 5 | 1355 | 85 | |
XM_017023033.1 | XP_016878522.1 | 4 - 5 | 532 | 85 | |
XM_017023034.1 | XP_016878523.1 | 3 - 4 | 451 | 85 | |
XM_017023035.1 | XP_016878524.1 | 3 - 4 | 480 | 85 | |
XM_017023036.1 | XP_016878525.1 | 2 - 3 | 1039 | 85 | |
XM_017023037.1 | XP_016878526.1 | 2 - 3 | 338 | 85 | |
XM_017023038.1 | XP_016878527.1 | 2 - 3 | 338 | 85 | |
XM_017023039.1 | XP_016878528.1 | 2 - 3 | 338 | 85 | |
GenBank mRNA | AK090431.1 | - | 2 - 3 | 392 | 85 |
AK090476.1 | - | 2 - 3 | 335 | 85 | |
AY601811.1 | - | 2 - 3 | 322 | 85 |