The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289004.1 | NP_001275933.1 | 2 - 3 | 784 | 114 |
NM_001308014.1 | NP_001294943.1 | 2 - 3 | 784 | 114 | |
GenBank mRNA | AK131445.1 | - | 2 - 3 | 681 | 114 |
BC034976.2 | - | 2 - 3 | 786 | 114 |