Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP1A1
Entrez Gene ID
1543
Gene Name
cytochrome P450 family 1 subfamily A member 1
Gene Aliases
AHH, AHRR, CP11, CYP1, CYPIA1, P1-450, P450-C, P450DX
Chromosome Location
Chr.15: 74719542 - 74725610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000499.4 NP_000490.1 4 - 5 1236 102
NM_001319216.1 NP_001306145.1 4 - 5 1239 102
NM_001319217.1 NP_001306146.1 4 - 5 1239 102
XM_017021953.1 XP_016877442.1 5 - 6 1155 102
GenBank mRNA AK223108.1 - 4 - 5 1157 102
AK223113.1 - 4 - 5 1154 102
AK310810.1 - 4 - 5 1283 102
AK313880.1 - 4 - 5 1154 102
AM233517.1 - 4 - 5 528 102
AM233518.1 - 4 - 5 528 102
AM233519.1 - 5 - 6 924 102
AM233520.1 - 5 - 6 1274 102
AM236046.1 - 5 - 6 1040 102
AM236047.1 - 4 - 5 335 102
AY310359.1 - 4 - 5 956 102
BC023019.1 - 4 - 5 1121 102
K03191.1 - 4 - 5 1126 102
M12079.1 - 2 - 3 158 102

View Full Product Details