The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178140.3 | NP_835260.2 | 8 - 9 | 2050 | 62 |
XM_005248269.4 | XP_005248326.1 | 8 - 9 | 2072 | 62 | |
XM_005248271.1 | XP_005248328.1 | 7 - 8 | 1424 | 62 | |
XM_005248272.3 | XP_005248329.1 | 7 - 8 | 1426 | 62 | |
XM_006714460.2 | XP_006714523.1 | 7 - 8 | 1223 | 62 | |
XM_011513992.2 | XP_011512294.1 | 9 - 10 | 2434 | 62 | |
XM_011513993.2 | XP_011512295.1 | 8 - 9 | 2352 | 62 | |
XM_011513994.2 | XP_011512296.1 | 9 - 10 | 2435 | 62 | |
XM_011513995.2 | XP_011512297.1 | 9 - 10 | 2433 | 62 | |
XM_011513996.2 | XP_011512298.1 | 7 - 8 | 2158 | 62 | |
XM_017009245.1 | XP_016864734.1 | 5 - 6 | 1148 | 62 | |
XM_017009246.1 | XP_016864735.1 | 9 - 10 | 2350 | 62 | |
GenBank mRNA | AB002298.2 | - | 8 - 9 | 2050 | 62 |
AK304804.1 | - | 7 - 8 | 1419 | 62 |