Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCND2
Entrez Gene ID
3751
Gene Name
potassium voltage-gated channel subfamily D member 2
Gene Aliases
KV4.2, RK5
Chromosome Location
Chr.7: 120273635 - 120750333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012281.2 NP_036413.1 1 - 2 2075 104
GenBank mRNA AB028967.1 - 1 - 2 2075 104
AJ010969.1 - 1 - 2 1539 104
BC110449.1 - 1 - 2 1729 104
BC110450.1 - 1 - 2 1729 104

View Full Product Details