Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAI
Entrez Gene ID
286205
Gene Name
suppressor of cancer cell invasion
Gene Aliases
C9orf126, NET40
Chromosome Location
Chr.9: 124942608 - 125143559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144877.2 NP_001138349.1 16 - 17 1700 74
NM_173690.4 NP_775961.2 17 - 18 1769 74
GenBank mRNA AF086497.1 - 3 - 4 314 74
AK093983.1 - 16 - 17 1659 74
AK098024.1 - 17 - 18 1706 74
BC104030.2 - 17 - 18 1697 74
BC104031.1 - 16 - 17 1628 74
BC104032.1 - 16 - 17 1546 74

View Full Product Details