The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024576.4 | NP_078852.3 | 3 - 4 | 897 | 160 |
XM_017011304.1 | XP_016866793.1 | 3 - 4 | 459 | 160 | |
GenBank mRNA | BC018688.1 | - | 3 - 4 | 532 | 160 |
BC110999.1 | - | 3 - 4 | 536 | 160 |