The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144881.1 | NP_001138353.1 | 7 - 8 | 1927 | 69 |
NM_001144882.1 | NP_001138354.1 | 7 - 8 | 1926 | 69 | |
NM_001144883.1 | NP_001138355.1 | 7 - 8 | 1923 | 69 | |
NM_153026.2 | NP_694571.2 | 7 - 8 | 1994 | 69 | |
XM_011537946.1 | XP_011536248.1 | 7 - 8 | 1745 | 69 | |
XM_011537947.2 | XP_011536249.1 | 7 - 8 | 1976 | 69 | |
XM_017018838.1 | XP_016874327.1 | 7 - 8 | 1790 | 69 | |
XM_017018839.1 | XP_016874328.1 | 7 - 8 | 1990 | 69 | |
XM_017018840.1 | XP_016874329.1 | 7 - 8 | 1810 | 69 | |
GenBank mRNA | AF399844.1 | - | 7 - 8 | 1900 | 69 |
AK056189.1 | - | 1 - 2 | 518 | 69 | |
AK056499.1 | - | 7 - 8 | 1900 | 69 | |
AK122877.1 | - | 7 - 8 | 1928 | 69 | |
AK122881.1 | - | 7 - 8 | 1871 | 69 | |
BC114939.1 | - | 7 - 8 | 1835 | 69 | |
BC114940.1 | - | 7 - 8 | 1836 | 69 |