The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012456.2 | NP_036588.1 | 1 - 2 | 99 | 70 |
GenBank mRNA | AF150089.1 | - | 1 - 2 | 81 | 70 |
AK289751.1 | - | 1 - 2 | 50 | 70 | |
BC032133.2 | - | 1 - 2 | 62 | 70 | |
CB106298.1 | - | 1 - 2 | 99 | 70 |