The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130517.2 | NP_001123989.1 | 1 - 2 | 135 | 83 |
GenBank mRNA | BX406051.2 | - | 1 - 2 | 81 | 83 |
DB450861.1 | - | 1 - 2 | 93 | 83 |