The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178335.2 | NP_848018.1 | 5 - 6 | 1043 | 71 |
XM_011512460.1 | XP_011510762.1 | 5 - 6 | 1046 | 71 | |
GenBank mRNA | AJ557013.1 | - | 5 - 6 | 1043 | 71 |