The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318792.1 | NP_001305721.1 | 16 - 17 | 1705 | 85 |
NM_018426.2 | NP_060896.1 | 16 - 17 | 1671 | 85 | |
XM_005249211.3 | XP_005249268.1 | 18 - 19 | 2051 | 85 | |
XM_005249213.4 | XP_005249270.1 | 17 - 18 | 1978 | 85 | |
XM_005249214.4 | XP_005249271.1 | 15 - 16 | 1565 | 85 | |
XM_005249215.4 | XP_005249272.1 | 16 - 17 | 1870 | 85 | |
XM_005249217.1 | XP_005249274.1 | 16 - 17 | 1743 | 85 | |
XM_006715135.1 | XP_006715198.1 | 16 - 17 | 1727 | 85 | |
XM_017010999.1 | XP_016866488.1 | 18 - 19 | 1928 | 85 | |
XM_017011000.1 | XP_016866489.1 | 17 - 18 | 1909 | 85 | |
XM_017011001.1 | XP_016866490.1 | 15 - 16 | 1609 | 85 | |
XM_017011002.1 | XP_016866491.1 | 15 - 16 | 1712 | 85 | |
GenBank mRNA | AL353957.1 | - | 2 - 3 | 129 | 85 |
AL834238.1 | - | 9 - 10 | 969 | 85 | |
BC022095.1 | - | 13 - 14 | 1308 | 85 | |
BC062989.1 | - | 4 - 5 | 497 | 85 | |
BC136769.1 | - | 15 - 16 | 1547 | 85 | |
CR627460.1 | - | 16 - 17 | 1708 | 85 |