Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
Chromosome Location
Chr.19: 47256474 - 47271958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015603.2 NP_056418.1 9 - 10 1151 87
XM_017026578.1 XP_016882067.1 9 - 10 1196 87
GenBank mRNA AL050284.1 - 9 - 10 1090 87
BC002787.2 - 9 - 10 1047 87
BC009743.2 - 9 - 10 1047 87

View Full Product Details