The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206855.1 | NP_001193784.1 | 7 - 8 | 1010 | 66 |
NM_003227.3 | NP_003218.2 | 10 - 11 | 1430 | 66 | |
XM_005250553.4 | XP_005250610.1 | 11 - 12 | 1736 | 66 | |
XM_017012573.1 | XP_016868062.1 | 11 - 12 | 1538 | 66 | |
GenBank mRNA | AF067864.1 | - | 10 - 11 | 1416 | 66 |
AK000421.1 | - | 6 - 7 | 742 | 66 | |
BC142630.1 | - | 10 - 11 | 1416 | 66 | |
BC150192.1 | - | 8 - 9 | 1137 | 66 |