Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location
Chr.8: 140658382 - 141001313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199649.1 NP_001186578.1 10 - 11 1174 85
NM_001316342.1 NP_001303271.1 8 - 9 918 85
NM_005607.4 NP_005598.3 10 - 11 1163 85
NM_153831.3 NP_722560.1 10 - 11 1174 85
XM_005251003.2 XP_005251060.1 9 - 10 1215 85
XM_005251012.2 XP_005251069.1 9 - 10 1057 85
XM_005251013.2 XP_005251070.1 8 - 9 876 85
XM_006716608.2 XP_006716671.1 10 - 11 1146 85
XM_006716609.2 XP_006716672.1 9 - 10 979 85
XM_006716610.2 XP_006716673.1 9 - 10 971 85
XM_011517194.2 XP_011515496.1 10 - 11 1115 85
XM_017013651.1 XP_016869140.1 8 - 9 896 85
XM_017013652.1 XP_016869141.1 8 - 9 896 85
XM_017013653.1 XP_016869142.1 9 - 10 1215 85
XM_017013654.1 XP_016869143.1 9 - 10 1215 85
XM_017013656.1 XP_016869145.1 9 - 10 1215 85
XM_017013657.1 XP_016869146.1 9 - 10 1215 85
XM_017013658.1 XP_016869147.1 10 - 11 1039 85
XM_017013659.1 XP_016869148.1 8 - 9 896 85
XM_017013660.1 XP_016869149.1 10 - 11 1039 85
XM_017013661.1 XP_016869150.1 9 - 10 1057 85
XM_017013662.1 XP_016869151.1 10 - 11 1039 85
XM_017013663.1 XP_016869152.1 9 - 10 970 85
XM_017013664.1 XP_016869153.1 10 - 11 1146 85
XM_017013665.1 XP_016869154.1 9 - 10 970 85
XM_017013669.1 XP_016869158.1 10 - 11 1039 85
XM_017013670.1 XP_016869159.1 10 - 11 1039 85
XM_017013671.1 XP_016869160.1 10 - 11 1039 85
XM_017013672.1 XP_016869161.1 9 - 10 970 85
XM_017013673.1 XP_016869162.1 9 - 10 970 85
XM_017013674.1 XP_016869163.1 10 - 11 1146 85
XM_017013675.1 XP_016869164.1 9 - 10 970 85
XM_017013676.1 XP_016869165.1 8 - 9 896 85
XM_017013680.1 XP_016869169.1 9 - 10 2031 85
XM_017013681.1 XP_016869170.1 9 - 10 979 85
XM_017013682.1 XP_016869171.1 8 - 9 876 85
XM_017013683.1 XP_016869172.1 9 - 10 979 85
XM_017013684.1 XP_016869173.1 9 - 10 979 85
XM_017013685.1 XP_016869174.1 9 - 10 923 85
XM_017013686.1 XP_016869175.1 8 - 9 876 85
XM_017013687.1 XP_016869176.1 9 - 10 979 85
GenBank mRNA AB209071.1 - 8 - 9 824 85
AB209083.1 - 10 - 11 761 85
AK304356.1 - 10 - 11 1019 85
AL832961.1 - 10 - 11 1046 85
BC035404.2 - 8 - 9 895 85
KF437463.1 - 8 - 9 867 85
L13616.1 - 10 - 11 1096 85

View Full Product Details