Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location
Chr.8: 140658382 - 141001313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199649.1 NP_001186578.1 19 - 20 1948 93
NM_001316342.1 NP_001303271.1 19 - 20 1731 93
NM_005607.4 NP_005598.3 19 - 20 1937 93
NM_153831.3 NP_722560.1 19 - 20 1948 93
XM_005251003.2 XP_005251060.1 21 - 22 2112 93
XM_005251012.2 XP_005251069.1 21 - 22 1954 93
XM_005251013.2 XP_005251070.1 20 - 21 1773 93
XM_006716608.2 XP_006716671.1 22 - 23 2043 93
XM_006716609.2 XP_006716672.1 21 - 22 1876 93
XM_006716610.2 XP_006716673.1 21 - 22 1868 93
XM_011517194.2 XP_011515496.1 22 - 23 2012 93
XM_017013651.1 XP_016869140.1 20 - 21 1793 93
XM_017013652.1 XP_016869141.1 20 - 21 1793 93
XM_017013653.1 XP_016869142.1 20 - 21 2028 93
XM_017013654.1 XP_016869143.1 19 - 20 2010 93
XM_017013655.1 XP_016869144.1 20 - 21 2004 93
XM_017013656.1 XP_016869145.1 19 - 20 2010 93
XM_017013657.1 XP_016869146.1 18 - 19 1989 93
XM_017013658.1 XP_016869147.1 21 - 22 1915 93
XM_017013659.1 XP_016869148.1 17 - 18 1670 93
XM_017013660.1 XP_016869149.1 21 - 22 1852 93
XM_017013661.1 XP_016869150.1 20 - 21 1870 93
XM_017013662.1 XP_016869151.1 20 - 21 1834 93
XM_017013663.1 XP_016869152.1 19 - 20 1765 93
XM_017013664.1 XP_016869153.1 20 - 21 1938 93
XM_017013665.1 XP_016869154.1 19 - 20 1762 93
XM_017013666.1 XP_016869155.1 17 - 18 1881 93
XM_017013667.1 XP_016869156.1 21 - 22 1828 93
XM_017013668.1 XP_016869157.1 20 - 21 1846 93
XM_017013669.1 XP_016869158.1 20 - 21 1834 93
XM_017013670.1 XP_016869159.1 20 - 21 1831 93
XM_017013671.1 XP_016869160.1 19 - 20 1813 93
XM_017013672.1 XP_016869161.1 18 - 19 1744 93
XM_017013673.1 XP_016869162.1 18 - 19 1744 93
XM_017013674.1 XP_016869163.1 21 - 22 1959 93
XM_017013675.1 XP_016869164.1 20 - 21 1783 93
XM_017013676.1 XP_016869165.1 17 - 18 1670 93
XM_017013677.1 XP_016869166.1 20 - 21 1744 93
XM_017013678.1 XP_016869167.1 17 - 18 1636 93
XM_017013679.1 XP_016869168.1 19 - 20 1675 93
XM_017013680.1 XP_016869169.1 21 - 22 2928 93
XM_017013681.1 XP_016869170.1 21 - 22 1876 93
XM_017013682.1 XP_016869171.1 19 - 20 1752 93
XM_017013683.1 XP_016869172.1 20 - 21 1792 93
XM_017013684.1 XP_016869173.1 19 - 20 1774 93
XM_017013685.1 XP_016869174.1 19 - 20 1715 93
XM_017013686.1 XP_016869175.1 17 - 18 1650 93
XM_017013687.1 XP_016869176.1 20 - 21 1792 93
XM_017013688.1 XP_016869177.1 19 - 20 1684 93
XM_017013689.1 XP_016869178.1 18 - 19 1549 93
XM_017013690.1 XP_016869179.1 17 - 18 1505 93
GenBank mRNA AB209071.1 - 19 - 20 1637 93
AB209083.1 - 22 - 23 1658 93
AK094999.1 - 9 - 10 654 93
AK294873.1 - 10 - 11 732 93
AK304356.1 - 19 - 20 1793 93
AL832961.1 - 19 - 20 1820 93
BC028733.2 - 5 - 6 596 93
BC035404.2 - 17 - 18 1669 93
KF437463.1 - 17 - 18 1641 93
L13616.1 - 19 - 20 1870 93

View Full Product Details