Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location
Chr.8: 140658382 - 141001313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316342.1 NP_001303271.1 28 - 29 2694 191
NM_005607.4 NP_005598.3 28 - 29 2900 191
NM_153831.3 NP_722560.1 28 - 29 2911 191
XM_005251003.2 XP_005251060.1 30 - 31 3075 191
XM_005251012.2 XP_005251069.1 30 - 31 2917 191
XM_005251013.2 XP_005251070.1 29 - 30 2736 191
XM_006716608.2 XP_006716671.1 31 - 32 3006 191
XM_006716609.2 XP_006716672.1 30 - 31 2839 191
XM_006716610.2 XP_006716673.1 30 - 31 2831 191
XM_011517194.2 XP_011515496.1 31 - 32 2975 191
XM_017013651.1 XP_016869140.1 29 - 30 2756 191
XM_017013652.1 XP_016869141.1 28 - 29 2693 128
XM_017013653.1 XP_016869142.1 29 - 30 2991 191
XM_017013654.1 XP_016869143.1 28 - 29 2973 191
XM_017013655.1 XP_016869144.1 29 - 30 2967 191
XM_017013656.1 XP_016869145.1 28 - 29 2973 191
XM_017013657.1 XP_016869146.1 27 - 28 2952 191
XM_017013658.1 XP_016869147.1 30 - 31 2878 191
XM_017013659.1 XP_016869148.1 25 - 26 2570 128
XM_017013660.1 XP_016869149.1 30 - 31 2815 191
XM_017013661.1 XP_016869150.1 29 - 30 2833 191
XM_017013662.1 XP_016869151.1 29 - 30 2797 191
XM_017013663.1 XP_016869152.1 28 - 29 2728 191
XM_017013664.1 XP_016869153.1 29 - 30 2901 191
XM_017013665.1 XP_016869154.1 28 - 29 2725 191
XM_017013666.1 XP_016869155.1 26 - 27 2844 191
XM_017013667.1 XP_016869156.1 30 - 31 2791 191
XM_017013668.1 XP_016869157.1 29 - 30 2809 191
XM_017013669.1 XP_016869158.1 29 - 30 2797 191
XM_017013670.1 XP_016869159.1 29 - 30 2794 191
XM_017013671.1 XP_016869160.1 28 - 29 2776 191
XM_017013672.1 XP_016869161.1 27 - 28 2707 191
XM_017013673.1 XP_016869162.1 27 - 28 2707 191
XM_017013674.1 XP_016869163.1 29 - 30 2859 128
XM_017013675.1 XP_016869164.1 28 - 29 2683 128
XM_017013676.1 XP_016869165.1 25 - 26 2570 128
XM_017013677.1 XP_016869166.1 29 - 30 2707 191
XM_017013678.1 XP_016869167.1 26 - 27 2599 191
XM_017013679.1 XP_016869168.1 27 - 28 2575 128
XM_017013680.1 XP_016869169.1 30 - 31 3891 191
XM_017013681.1 XP_016869170.1 30 - 31 2839 191
XM_017013682.1 XP_016869171.1 28 - 29 2715 191
XM_017013683.1 XP_016869172.1 29 - 30 2755 191
XM_017013684.1 XP_016869173.1 28 - 29 2737 191
XM_017013685.1 XP_016869174.1 28 - 29 2678 191
XM_017013686.1 XP_016869175.1 26 - 27 2613 191
XM_017013687.1 XP_016869176.1 28 - 29 2692 128
XM_017013688.1 XP_016869177.1 28 - 29 2647 191
XM_017013689.1 XP_016869178.1 27 - 28 2512 191
XM_017013690.1 XP_016869179.1 26 - 27 2468 191
GenBank mRNA AB209071.1 - 28 - 29 2600 191
AB209083.1 - 31 - 32 2621 191
AK094999.1 - 18 - 19 1617 191
AK294873.1 - 19 - 20 1695 191
AK301557.1 - 7 - 8 616 191
AL832961.1 - 28 - 29 2783 191
BC028733.2 - 14 - 15 1559 191
BC035404.2 - 25 - 26 2494 191
KF437463.1 - 26 - 27 2604 191
L13616.1 - 28 - 29 2833 191

View Full Product Details