Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
Chromosome Location
Chr.7: 87345635 - 87399796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143935.1 NP_001137407.1 17 - 18 1928 86
NM_021151.3 NP_066974.2 16 - 17 1844 86
XM_011516337.2 XP_011514639.1 16 - 17 1844 86
XM_017012370.1 XP_016867859.1 17 - 18 1918 86
GenBank mRNA AF052187.1 - 2 - 3 115 86
AF073770.1 - 16 - 17 1735 86
AF168793.1 - 16 - 17 1735 86
AK127901.1 - 17 - 18 1884 86
AK300114.1 - 11 - 12 1174 86
AK313487.1 - 16 - 17 1787 86
BC039004.1 - 16 - 17 1784 86

View Full Product Details