Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
Chromosome Location
Chr.7: 87345635 - 87399796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143935.1 NP_001137407.1 18 - 19 2043 92
NM_021151.3 NP_066974.2 17 - 18 1959 92
XM_017012370.1 XP_016867859.1 18 - 19 2033 92
GenBank mRNA AF052187.1 - 3 - 4 230 92
AF073770.1 - 17 - 18 1850 92
AF168793.1 - 17 - 18 1850 92
AK127901.1 - 18 - 19 1999 92
AK300114.1 - 12 - 13 1289 92
AK313487.1 - 17 - 18 1902 92
BC039004.1 - 17 - 18 1899 92

View Full Product Details