The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013448.2 | NP_038476.2 | 2 - 3 | 676 | 86 |
NM_182648.1 | NP_872589.1 | 2 - 3 | 676 | 86 | |
GenBank mRNA | AB032252.1 | - | 2 - 3 | 673 | 86 |
AF086397.1 | - | 2 - 3 | 274 | 86 | |
AF161435.1 | - | 2 - 3 | 362 | 86 | |
AF213467.1 | - | 2 - 3 | 274 | 86 | |
AF221130.1 | - | 1 - 2 | 108 | 86 | |
BC020636.1 | - | 1 - 2 | 436 | 86 | |
BC108711.1 | - | 2 - 3 | 340 | 86 |