Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPM1A
- Entrez Gene ID
-
5494
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1A
- Gene Aliases
-
PP2C-ALPHA, PP2CA, PP2Calpha
- Chromosome Location
-
Chr.14: 60245752 - 60299087 on Build GRCh38
View Full Product Details