Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
Chromosome Location
Chr.16: 81314941 - 81380198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022041.3 NP_071324.1 10 - 11 1754 101
XM_017023734.1 XP_016879223.1 9 - 10 1664 101
GenBank mRNA AF291673.1 - 10 - 11 1750 101
AK095378.1 - 10 - 11 1754 101
BC044840.1 - 10 - 11 1741 101
KU178409.1 - 10 - 11 1606 101
KU178410.1 - 8 - 9 1140 101

View Full Product Details