The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 15 - 16 | 1636 | 62 |
NM_001193315.1 | NP_001180244.1 | 15 - 16 | 1236 | 62 | |
NM_001193316.1 | NP_001180245.1 | 14 - 15 | 1089 | 62 | |
NM_001193317.1 | NP_001180246.1 | 15 - 16 | 1413 | 62 | |
NM_022067.3 | NP_071350.2 | 16 - 17 | 1507 | 62 | |
XM_011537066.1 | XP_011535368.1 | 14 - 15 | 1134 | 62 | |
XM_017021579.1 | XP_016877068.1 | 14 - 15 | 1268 | 62 | |
XM_017021580.1 | XP_016877069.1 | 15 - 16 | 1227 | 62 | |
XM_017021581.1 | XP_016877070.1 | 15 - 16 | 1361 | 62 | |
GenBank mRNA | AK022769.1 | - | 15 - 16 | 1184 | 62 |
AK022925.1 | - | 15 - 16 | 1162 | 62 | |
AK024661.1 | - | 4 - 5 | 332 | 62 | |
AK098254.1 | - | 15 - 16 | 1597 | 62 | |
AK222706.1 | - | 15 - 16 | 1182 | 62 | |
AK297388.1 | - | 15 - 16 | 1338 | 62 | |
AK298354.1 | - | 14 - 15 | 1035 | 62 | |
BC015054.1 | - | 16 - 17 | 1428 | 62 | |
CR457294.1 | - | 14 - 15 | 1090 | 62 |