The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 19 - 20 | 2011 | 81 |
NM_001193315.1 | NP_001180244.1 | 19 - 20 | 1611 | 81 | |
NM_001193316.1 | NP_001180245.1 | 18 - 19 | 1464 | 81 | |
NM_001193317.1 | NP_001180246.1 | 19 - 20 | 1788 | 81 | |
NM_022067.3 | NP_071350.2 | 20 - 21 | 1882 | 81 | |
XM_011537066.1 | XP_011535368.1 | 18 - 19 | 1509 | 81 | |
XM_017021579.1 | XP_016877068.1 | 18 - 19 | 1643 | 81 | |
GenBank mRNA | AK022769.1 | - | 19 - 20 | 1559 | 81 |
AK022925.1 | - | 19 - 20 | 1537 | 81 | |
AK024661.1 | - | 8 - 9 | 707 | 81 | |
AK098254.1 | - | 19 - 20 | 1972 | 81 | |
AK297388.1 | - | 19 - 20 | 1713 | 81 | |
AK298354.1 | - | 18 - 19 | 1410 | 81 | |
BC015054.1 | - | 20 - 21 | 1803 | 81 |