The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318508.1 | NP_001305437.1 | 6 - 7 | 541 | 79 |
NM_015962.4 | NP_057046.1 | 6 - 7 | 505 | 79 | |
XM_005267730.3 | XP_005267787.1 | 6 - 7 | 637 | 79 | |
XM_011536815.2 | XP_011535117.1 | 5 - 6 | 462 | 79 | |
XM_011536816.2 | XP_011535118.1 | 5 - 6 | 462 | 79 | |
GenBank mRNA | AF132969.1 | - | 5 - 6 | 454 | 79 |
AK290666.1 | - | 6 - 7 | 486 | 79 | |
BC080600.1 | - | 6 - 7 | 465 | 79 | |
BX248271.1 | - | 6 - 7 | 467 | 79 | |
BX489015.1 | - | 6 - 7 | 507 | 79 | |
BX647810.1 | - | 6 - 7 | 531 | 79 |