The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003659.3 | NP_003650.1 | 12 - 13 | 1332 | 149 |
XM_011512041.2 | XP_011510343.1 | 12 - 13 | 1114 | 149 | |
XM_011512042.1 | XP_011510344.1 | 12 - 13 | 1281 | 149 | |
XM_011512043.2 | XP_011510345.1 | 8 - 9 | 1918 | 149 | |
GenBank mRNA | AK296239.1 | - | 12 - 13 | 1112 | 149 |
AK309714.1 | - | 12 - 13 | 1431 | 149 | |
AK314259.1 | - | 12 - 13 | 1302 | 149 | |
AK316240.1 | - | 12 - 13 | 1113 | 149 | |
AY544121.1 | - | 12 - 13 | 1285 | 149 | |
BC141820.1 | - | 12 - 13 | 1297 | 149 | |
Y09443.1 | - | 12 - 13 | 1300 | 149 |