The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003036.3 | NP_003027.1 | 2 - 3 | 1172 | 50 |
XM_005244775.3 | XP_005244832.1 | 2 - 3 | 1142 | 50 | |
XM_005244776.4 | XP_005244833.1 | 2 - 3 | 4285 | 50 | |
XM_017002128.1 | XP_016857617.1 | 2 - 3 | 949 | 50 | |
GenBank mRNA | X15218.1 | - | 2 - 3 | 1172 | 50 |