Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNHIT2
Entrez Gene ID
741
Gene Name
zinc finger HIT-type containing 2
Gene Aliases
C11orf5, FON
Chromosome Location
Chr.11: 65116403 - 65117738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014205.3 NP_055020.1 1 - 1 1061 114
GenBank mRNA AF119497.1 - 1 - 1 1021 114
AK027081.1 - 1 - 1 996 114
AK223333.1 - 1 - 1 976 114
AK223358.1 - 1 - 1 1031 114
BC052240.1 - 1 - 1 1024 114
BC104002.2 - 1 - 1 114
BC104003.2 - 1 - 1 114
HY200798.1 - 1 - 1 114

View Full Product Details