Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DPP8
- Entrez Gene ID
-
54878
- Gene Name
- dipeptidyl peptidase 8
- Gene Aliases
-
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
- Chromosome Location
-
Chr.15: 65442463 - 65517713 on Build GRCh38
View Full Product Details