Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF1B
Entrez Gene ID
9014
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit B
Gene Aliases
MGC:9349, RAF1B, RAFI63, SL1, TAFI63
Chromosome Location
Chr.2: 9843442 - 9934416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005680.2 NP_005671.2 7 - 8 802 129
XM_011510410.2 XP_011508712.1 7 - 8 743 129
XM_011510411.1 XP_011508713.1 8 - 9 851 129
GenBank mRNA AK308070.1 - 7 - 8 751 129
AK309751.1 - 7 - 8 785 129
AK315098.1 - 7 - 8 734 129
BC018137.2 - 7 - 8 801 129
L39061.1 - 7 - 8 804 129

View Full Product Details