Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
Chromosome Location
Chr.1: 62236165 - 62319414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320269.1 NP_001307198.1 7 - 8 1176 92
NM_181712.4 NP_859063.3 8 - 9 3058 92
XM_011540843.2 XP_011539145.1 8 - 9 2943 92
XM_011540845.2 XP_011539147.1 8 - 9 2937 92
XM_017000484.1 XP_016855973.1 8 - 9 1312 92
XM_017000485.1 XP_016855974.1 6 - 7 1244 92
GenBank mRNA AB284127.1 - 8 - 9 3058 92
AK091941.1 - 7 - 8 1063 92
AK096259.1 - 2 - 3 442 92
AK127461.1 - 5 - 6 1984 92
AL832644.2 - 6 - 7 1810 92
BC041418.1 - 6 - 7 1278 92
BC060866.1 - 8 - 9 3058 92

View Full Product Details