The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034023.1 | NP_001029195.1 | 9 - 10 | 1178 | 58 |
NM_001034024.1 | NP_001029196.1 | 9 - 10 | 1138 | 58 | |
NM_019100.4 | NP_061973.1 | 8 - 9 | 1359 | 58 | |
XM_005271037.4 | XP_005271094.1 | 8 - 9 | 1349 | 58 | |
XM_005271039.1 | XP_005271096.1 | 9 - 10 | 1249 | 58 | |
XM_005271040.1 | XP_005271097.1 | 9 - 10 | 1126 | 58 | |
XM_006710771.3 | XP_006710834.1 | 7 - 8 | 1028 | 58 | |
XM_011541784.2 | XP_011540086.1 | 9 - 10 | 1183 | 58 | |
XM_011541785.2 | XP_011540087.1 | 9 - 10 | 1203 | 58 | |
XM_017001807.1 | XP_016857296.1 | 9 - 10 | 1234 | 58 | |
XM_017001808.1 | XP_016857297.1 | 9 - 10 | 1237 | 58 | |
GenBank mRNA | AB037846.1 | - | 9 - 10 | 1143 | 58 |
AF265228.1 | - | 8 - 9 | 1058 | 58 | |
AK021605.1 | - | 9 - 10 | 1122 | 58 | |
AK054954.1 | - | 5 - 6 | 1976 | 58 | |
AK092898.1 | - | 5 - 6 | 4143 | 58 | |
AK096252.1 | - | 5 - 6 | 2746 | 58 | |
AK289366.1 | - | 9 - 10 | 1161 | 58 | |
AK298793.1 | - | 9 - 10 | 1121 | 58 | |
AL136657.1 | - | 9 - 10 | 1140 | 58 | |
AL137200.1 | - | 9 - 10 | 1142 | 58 | |
BC002855.2 | - | 9 - 10 | 1127 | 58 | |
BC008053.2 | - | 8 - 9 | 1321 | 58 | |
BU616029.1 | - | 2 - 3 | 58 | ||
BX537895.1 | - | 5 - 6 | 3297 | 58 |