The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032839.2 | NP_116228.1 | 7 - 8 | 1278 | 72 |
GenBank mRNA | AK027690.1 | - | 7 - 8 | 1272 | 72 |
AK291176.1 | - | 7 - 8 | 1262 | 72 | |
BC022204.1 | - | 7 - 8 | 1536 | 72 | |
BC033738.1 | - | 6 - 7 | 1154 | 72 | |
BC039821.1 | - | 7 - 8 | 1249 | 72 |