The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016519.5 | NP_057603.1 | 7 - 8 | 665 | 75 |
GenBank mRNA | AF219994.2 | - | 7 - 8 | 629 | 75 |
AF263464.1 | - | 7 - 8 | 639 | 75 | |
AJ537437.1 | - | 7 - 8 | 639 | 75 | |
AK314918.1 | - | 7 - 8 | 600 | 75 | |
BC106931.1 | - | 7 - 8 | 600 | 75 |