The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006982.2 | NP_008913.2 | 3 - 4 | 659 | 91 |
XM_011538782.2 | XP_011537084.1 | 3 - 4 | 684 | 91 | |
GenBank mRNA | BC010923.1 | - | 3 - 4 | 665 | 91 |
EU794597.1 | - | 3 - 4 | 665 | 91 | |
U31986.1 | - | 3 - 4 | 810 | 91 |