The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304504.1 | NP_001291433.1 | 8 - 9 | 1258 | 110 |
NM_001304505.1 | NP_001291434.1 | 8 - 9 | 1170 | 110 | |
NM_018200.3 | NP_060670.1 | 9 - 10 | 1351 | 110 | |
XM_011521158.2 | XP_011519460.1 | 8 - 9 | 1214 | 110 | |
GenBank mRNA | AF146222.1 | - | 8 - 9 | 991 | 110 |
AK001601.1 | - | 8 - 9 | 1021 | 110 | |
AK223100.1 | - | 8 - 9 | 1017 | 110 | |
AK297449.1 | - | 7 - 8 | 951 | 110 | |
AK316281.1 | - | 5 - 6 | 626 | 110 | |
AL355737.1 | - | 7 - 8 | 913 | 110 | |
BC021959.1 | - | 9 - 10 | 1101 | 110 | |
BT006716.1 | - | 7 - 8 | 900 | 110 |