The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004528.3 | NP_004519.1 | 2 - 3 | 541 | 102 |
XM_005245174.3 | XP_005245231.2 | 2 - 3 | 144 | 102 | |
GenBank mRNA | AF026977.1 | - | 1 - 2 | 162 | 102 |
AK223012.1 | - | 1 - 2 | 141 | 102 | |
AK312103.1 | - | 2 - 3 | 157 | 102 | |
BC000505.2 | - | 2 - 3 | 140 | 102 | |
BC003034.2 | - | 1 - 2 | 122 | 102 | |
BC005964.1 | - | 2 - 3 | 190 | 102 | |
BI666647.1 | - | 2 - 3 | 206 | 102 | |
CR450359.1 | - | 1 - 2 | 113 | 102 |