Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL2
Entrez Gene ID
10150
Gene Name
muscleblind like splicing regulator 2
Gene Aliases
MBLL, MBLL39, PRO2032
Chromosome Location
Chr.13: 97141780 - 97394120 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306070.1 NP_001292999.1 5 - 6 1650 140
NM_144778.3 NP_659002.1 5 - 6 1643 140
NM_207304.2 NP_997187.1 5 - 6 1643 140
XM_005254022.2 XP_005254079.1 5 - 6 1928 140
XM_017020309.1 XP_016875798.1 5 - 6 1519 140
XM_017020310.1 XP_016875799.1 5 - 6 1481 140
XM_017020311.1 XP_016875800.1 5 - 6 1519 140
GenBank mRNA AF491866.1 - 5 - 6 1584 140
AK291727.1 - 5 - 6 1590 140
AK315844.1 - 5 - 6 1566 140
AY101770.1 - 4 - 5 878 140
BC104038.1 - 5 - 6 1571 140
BC104039.1 - 5 - 6 1571 140
BC104040.1 - 5 - 6 1571 140
CR749802.1 - 6 - 7 1602 140

View Full Product Details