Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARP15
Entrez Gene ID
165631
Gene Name
poly(ADP-ribose) polymerase family member 15
Gene Aliases
ARTD7, BAL3, pART7
Chromosome Location
Chr.3: 122575914 - 122639047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113523.2 NP_001106995.1 4 - 5 834 108
XM_005247159.3 XP_005247216.1 4 - 5 2506 108
XM_011512475.2 XP_011510777.1 3 - 4 2390 108
XM_011512480.2 XP_011510782.1 4 - 5 2522 108
GenBank mRNA DQ063586.1 - 4 - 5 753 108

View Full Product Details