The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152305.2 | NP_689518.1 | 4 - 5 | 544 | 74 |
GenBank mRNA | AF168711.1 | - | 4 - 5 | 495 | 74 |
AK075444.1 | - | 4 - 5 | 480 | 74 | |
AK222937.1 | - | 4 - 5 | 504 | 74 | |
AK315367.1 | - | 4 - 5 | 519 | 74 | |
AY298903.1 | - | 4 - 5 | 530 | 74 | |
AY358581.1 | - | 4 - 5 | 487 | 74 | |
BC030614.2 | - | 4 - 5 | 530 | 74 | |
BC048810.1 | - | 4 - 5 | 488 | 74 |